Learn More
Medchemexpress LLC Azido-PEG3-O-NHS ester | 2110448-98-9 | 95% | C13H20N4O8 | 100 MG
SDP
Catalog No. 5000353522
Encompass_Preferred
Azido-PEG3-O-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent features an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN group-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-53522 Supplier Medchemexpress LLC Supplier No. HY140768100MG
Azido-PEG3-O-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent features an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN group-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups