Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG3-propionic acid tert-butyl ester | 252881-73-5 | MFCD22201543 | ≥98.0% | 303.35 g/mol | C13H25N3O5 | 100 MG
SDP

Catalog No. 5000203363
Encompass_Preferred

Azido-PEG3-propionic acid tert-butyl ester is an azide-functionalized, Boc-protected PEG3 linker for bioconjugation. It acts as a cleavable three-unit PEG spacer and an azide reagent for click chemistry, suitable for assembling antibody-drug conjugates and PROTACs.

  • Azide-functionalized Boc-protected PEG3 linker.
  • Cleavable three-unit PEG spacer for ADC and PROTAC synthesis.
  • Suitable for CuAAC and SPAAC click chemistry.
  • Purity ≥98.0%.
  • Molecular weight 303.35 g/mol; formula C13H25N3O5.
  • Appearance colorless to light yellow liquid; follow storage recommendations.

Catalog No. 50-002-03363 Supplier Medchemexpress LLC Supplier No. HY42489100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG3-propionic acid tert-butyl ester is an azide-functionalized, Boc-protected PEG3 linker for bioconjugation. It acts as a cleavable three-unit PEG spacer and an azide reagent for click chemistry, suitable for assembling antibody-drug conjugates and PROTACs.

  • Azide-functionalized Boc-protected PEG3 linker.
  • Cleavable three-unit PEG spacer for ADC and PROTAC synthesis.
  • Suitable for CuAAC and SPAAC click chemistry.
  • Purity ≥98.0%.
  • Molecular weight 303.35 g/mol; formula C13H25N3O5.
  • Appearance colorless to light yellow liquid; follow storage recommendations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.