Learn More
Medchemexpress LLC Azido-PEG3-SS-NHS | 436.50 | C15H24N4O7S2 | 10 MG
SDP
Catalog No. 5000202402
Encompass_Preferred
Azido-PEG3-SS-NHS is an azide-functionalized, cleavable PEG3 linker used for bioconjugation and antibody-drug conjugate (ADC) synthesis. It contains an NHS ester for amine coupling, a disulfide (SS) cleavage site, and an azide group for click chemistry reactions (CuAAC and SPAAC).
  • Cleavable disulfide linker enabling reductive release
  • Contains NHS ester for efficient amine conjugation
  • Azide group supports CuAAC and strain-promoted click chemistries
  • Liquid form; molecular weight 436.50 and formula C15H24N4O7S2
  • Available in small milligram quantities (e.g., 5 mg, 10 mg)
Catalog No. 50-002-02402 Supplier Medchemexpress LLC Supplier No. HY13596610MG
Azido-PEG3-SS-NHS is an azide-functionalized, cleavable PEG3 linker used for bioconjugation and antibody-drug conjugate (ADC) synthesis. It contains an NHS ester for amine coupling, a disulfide (SS) cleavage site, and an azide group for click chemistry reactions (CuAAC and SPAAC).
  • Cleavable disulfide linker enabling reductive release
  • Contains NHS ester for efficient amine conjugation
  • Azide group supports CuAAC and strain-promoted click chemistries
  • Liquid form; molecular weight 436.50 and formula C15H24N4O7S2
  • Available in small milligram quantities (e.g., 5 mg, 10 mg)