Learn More
Medchemexpress LLC Azido-PEG3-SS-NHS | 95.0% | 436.50 | C15H24N4O7S2 | 5 MG
SDP
Catalog No. 5000202399
Encompass_Preferred
Azido-PEG3-SS-NHS is a cleavable three-unit polyethylene glycol (PEG) linker intended for bioconjugation and antibody-drug conjugate synthesis. It contains an azide for click chemistry, a disulfide bond for reductive cleavage, and an N-hydroxysuccinimide (NHS) ester for amine coupling.
  • Contains azide for copper-catalyzed and strain-promoted click reactions.
  • Contains NHS ester for efficient conjugation to primary amines.
  • Includes a cleavable disulfide spacer for intracellular release.
  • Suitable for linker design in ADC, PROTAC, and other bioconjugation workflows.
  • Supplied as a liquid, colorless to light yellow, with reported purity of 95.0%.
Catalog No. 50-002-02399 Supplier Medchemexpress LLC Supplier No. HY1359665MG
Azido-PEG3-SS-NHS is a cleavable three-unit polyethylene glycol (PEG) linker intended for bioconjugation and antibody-drug conjugate synthesis. It contains an azide for click chemistry, a disulfide bond for reductive cleavage, and an N-hydroxysuccinimide (NHS) ester for amine coupling.
  • Contains azide for copper-catalyzed and strain-promoted click reactions.
  • Contains NHS ester for efficient conjugation to primary amines.
  • Includes a cleavable disulfide spacer for intracellular release.
  • Suitable for linker design in ADC, PROTAC, and other bioconjugation workflows.
  • Supplied as a liquid, colorless to light yellow, with reported purity of 95.0%.