Learn More
Medchemexpress LLC Azido-PEG3-SS-PEG3-azide | 1310827-27-0 | 99.3% | 468.59 | 50 MG
SDP
Catalog No. 5000349206
Encompass_Preferred
Azido-PEG3-SS-PEG3-azide is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs themselves are agents that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • A PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains an Azide group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Alkyne-containing molecules.
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN-containing molecules.
Catalog No. 50-003-49206 Supplier Medchemexpress LLC Supplier No. HY14010650MG
Azido-PEG3-SS-PEG3-azide is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs themselves are agents that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • A PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains an Azide group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Alkyne-containing molecules.
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN-containing molecules.