Learn More
Medchemexpress LLC Azido-PEG3-SS-PEG3-azide | 1310827-27-0 | 99.3% | C16H32N6O6S2 | 10MG
SDP
Catalog No. 5000260194
Encompass_Preferred
Azido-PEG3-SS-PEG3-azide is a symmetric, cleavable polyethylene glycol (PEG)-based bifunctional linker bearing terminal azide groups and a central disulfide bond. It is designed for bioconjugation and PROTAC synthesis, enabling both copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), while allowing reductive cleavage for controlled release.
Catalog No. 50-002-60194 Supplier Medchemexpress LLC Supplier No. HY14010610MG
Azido-PEG3-SS-PEG3-azide is a symmetric, cleavable polyethylene glycol (PEG)-based bifunctional linker bearing terminal azide groups and a central disulfide bond. It is designed for bioconjugation and PROTAC synthesis, enabling both copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), while allowing reductive cleavage for controlled release.