Learn More
Medchemexpress LLC Azido-PEG3-SS-PEG3-azide | 1310827-27-0 | MFCD28976667 | 99.3% | 468.59 | 25 MG
SDP
Catalog No. 5000349054
Encompass_Preferred
Azido-PEG3-SS-PEG3-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent containing an azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent containing an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-49054 Supplier Medchemexpress LLC Supplier No. 25MGHY140106
Azido-PEG3-SS-PEG3-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent containing an azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent containing an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups