Learn More
Medchemexpress LLC Azido-PEG3-SSPy | 98.6% | 344.45 g·mol⁻¹ | C13H20N4O3S2 | 100 MG
SDP
Catalog No. 5000203821
Encompass_Preferred
Azido-PEG3-SSPy is a cleavable three-unit polyethylene glycol (PEG3) linker used for assembling antibody-drug conjugates (ADCs) and general bioconjugation. It contains an azide functional group enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) and is compatible with strain-promoted azide-alkyne cycloaddition (SPAAC) with DBCO or BCN partners.
  • Cleavable disulfide linker for controlled release.
  • Azide-functionalized for CuAAC click chemistry.
  • Compatible with SPAAC (DBCO and BCN) reactions.
  • High purity suitable for conjugation workflows.
  • Multiple pack sizes available for research use.
Catalog No. 50-002-03821 Supplier Medchemexpress LLC Supplier No. HY136038100MG
Azido-PEG3-SSPy is a cleavable three-unit polyethylene glycol (PEG3) linker used for assembling antibody-drug conjugates (ADCs) and general bioconjugation. It contains an azide functional group enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) and is compatible with strain-promoted azide-alkyne cycloaddition (SPAAC) with DBCO or BCN partners.
  • Cleavable disulfide linker for controlled release.
  • Azide-functionalized for CuAAC click chemistry.
  • Compatible with SPAAC (DBCO and BCN) reactions.
  • High purity suitable for conjugation workflows.
  • Multiple pack sizes available for research use.