Learn More
Medchemexpress LLC Azido-PEG3-SSPy | 98.6% | 344.45 g/mol | C13H20N4O3S2 | 50 MG
SDP
Catalog No. 5000203948
Encompass_Preferred
Azido-PEG3-SSPy is a cleavable three-unit polyethylene glycol (PEG3) linker that contains an azide functional group and a disulfide-pyridyldithiol (SSPy) moiety; it is designed for bioconjugation and ADC linker synthesis using click chemistry.
  • Cleavable PEG3 linker enabling disulfide-mediated release.
  • Azide functional group supports CuAAC and SPAAC bioconjugation.
  • High purity (98.6%) suitable for research applications.
  • Molecular weight 344.45 g/mol and formula C13H20N4O3S2.
  • Available as a 50 MG vial and stored per manufacturer recommendations.
Catalog No. 50-002-03948 Supplier Medchemexpress LLC Supplier No. HY13603850MG
Azido-PEG3-SSPy is a cleavable three-unit polyethylene glycol (PEG3) linker that contains an azide functional group and a disulfide-pyridyldithiol (SSPy) moiety; it is designed for bioconjugation and ADC linker synthesis using click chemistry.
  • Cleavable PEG3 linker enabling disulfide-mediated release.
  • Azide functional group supports CuAAC and SPAAC bioconjugation.
  • High purity (98.6%) suitable for research applications.
  • Molecular weight 344.45 g/mol and formula C13H20N4O3S2.
  • Available as a 50 MG vial and stored per manufacturer recommendations.