Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azido-PEG3-sulforhodamine 101 conjugate, Thermo Scientific Chemicals

Catalog No. AAJ65476LB0
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65476LB0 5 mg
AAJ65476MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65476LB0 Supplier Thermo Scientific Chemicals Supplier No. J65476LB0
Add to Cart

A water soluble sulforhodamine 101 with azido group functionality

A water soluble sulforhodamine 101 with azido group functionality

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1353153-76-0
Molecular Formula C39H46N6O9S2
MDL Number MFCD22380752
Synonym Azide-Fluor 585

Specifications

Color Red
Quantity 5 mg
Formula Weight 806.95
Physical Form Solid
Chemical Name or Material Azido-PEG3-sulforhodamine 101 conjugate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.