Learn More
Medchemexpress LLC Azido-PEG3-Val-Cit-PAB-PNP | 2055047-18-0 | 99.1% | 773.79 g/mol | C34H47N9O12 | 100 MG
SDP
Catalog No. 5000203005
Encompass_Preferred
Azido-PEG3-Val-Cit-PAB-PNP is an azide-functional PEG3 Val-Cit PAB cleavable linker designed for use in antibody-drug conjugate (ADC) assembly and in the construction of PROTAC linkers. It combines an azide handle for click chemistry with a PNP-activated PAB leaving group for efficient conjugation.
  • Cleavable Val-Cit PAB dipeptide linker suitable for ADCs.
  • Azide-functionalized for copper-catalyzed and strain-promoted click reactions.
  • PEG3 spacer to improve hydrophilicity and linker flexibility.
  • PNP-activated leaving group for efficient conjugation chemistry.
  • Available in small-scale quantities including 100 mg for research use.
Catalog No. 50-002-03005 Supplier Medchemexpress LLC Supplier No. HY140150100MG
Azido-PEG3-Val-Cit-PAB-PNP is an azide-functional PEG3 Val-Cit PAB cleavable linker designed for use in antibody-drug conjugate (ADC) assembly and in the construction of PROTAC linkers. It combines an azide handle for click chemistry with a PNP-activated PAB leaving group for efficient conjugation.
  • Cleavable Val-Cit PAB dipeptide linker suitable for ADCs.
  • Azide-functionalized for copper-catalyzed and strain-promoted click reactions.
  • PEG3 spacer to improve hydrophilicity and linker flexibility.
  • PNP-activated leaving group for efficient conjugation chemistry.
  • Available in small-scale quantities including 100 mg for research use.