Learn More
Medchemexpress LLC Azido-PEG4-alcohol | 86770-67-4 | 98.88% | 219.24 | 1 G
SDP
Catalog No. 5000400685
Encompass_Preferred
This is a PEG-based linker used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules that have Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based linker
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-004-00685 Supplier Medchemexpress LLC Supplier No. HY223401G
This is a PEG-based linker used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules that have Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based linker
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions