Learn More
Medchemexpress LLC Azido-PEG4-alcohol | 86770-67-4 | 98.9% | 219.24 | 5 G
SDP
Catalog No. 5000400820
Encompass_Preferred
Azido-PEG4-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, able to undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. PROTACs themselves contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-004-00820 Supplier Medchemexpress LLC Supplier No. HY223405G
Azido-PEG4-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, able to undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. PROTACs themselves contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions