Learn More
Medchemexpress LLC Azido-PEG4-alpha-D-mannose | 1632372-86-1 | 96.2% | C14H27N3O9 | 10MG
SDP
Catalog No. 5000260241
Encompass_Preferred
Azido-PEG4-alpha-D-mannose is a PEG4 linker with an azide functional group and an alpha-D-mannose residue, used for click-chemistry conjugation and glyco-conjugation in chemical biology and bioconjugation workflows.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for efficient bioconjugation.
  • Provides a short, hydrophilic PEG4 spacer to improve solubility and reduce steric hindrance.
  • Presents an alpha-D-mannose moiety for targeting mannose-binding lectins and receptors.
  • Reported purity around 96.2% suitable for research applications.
  • Available in small pack sizes for synthetic and analytical workflows.
Catalog No. 50-002-60241 Supplier Medchemexpress LLC Supplier No. HY14112610MG
Azido-PEG4-alpha-D-mannose is a PEG4 linker with an azide functional group and an alpha-D-mannose residue, used for click-chemistry conjugation and glyco-conjugation in chemical biology and bioconjugation workflows.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for efficient bioconjugation.
  • Provides a short, hydrophilic PEG4 spacer to improve solubility and reduce steric hindrance.
  • Presents an alpha-D-mannose moiety for targeting mannose-binding lectins and receptors.
  • Reported purity around 96.2% suitable for research applications.
  • Available in small pack sizes for synthetic and analytical workflows.