Learn More
Medchemexpress LLC Azido-PEG4-Amido-tri-(t-butoxycarbonylethoxymethyl)-methane | 1421933-29-0 | 97.0% | C36H66N4O14 | 25 MG
SDP
Catalog No. 5000352635
Encompass_Preferred
This compound is a PEG-based PROTAC linker, instrumental in the synthesis of PROTACs. It also serves as a click chemistry reagent, featuring an azide group. This enables it to engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. PROTACs leverage an intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Click chemistry reagent with an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Exploits the ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-52635 Supplier Medchemexpress LLC Supplier No. 25MGHY140874
This compound is a PEG-based PROTAC linker, instrumental in the synthesis of PROTACs. It also serves as a click chemistry reagent, featuring an azide group. This enables it to engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. PROTACs leverage an intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Click chemistry reagent with an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Exploits the ubiquitin-proteasome system for selective protein degradation