Learn More
Medchemexpress LLC Azido-PEG4-Amido-tri-(t-butoxycarbonylethoxymethyl)-methane | 1421933-29-0 | 97.0% | C36H66N4O14 | 50 MG
SDP
Catalog No. 5000355109
Encompass_Preferred
Azido-PEG4-Amido-tri-(t-butoxycarbonylethoxymethyl)-methane is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PROTAC linker
  • Click chemistry reagent
  • Contains an azide group for CuAAc reaction
  • Can undergo SPAAC reactions with DBCO or BCN groups
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-55109 Supplier Medchemexpress LLC Supplier No. HY14087450MG
Azido-PEG4-Amido-tri-(t-butoxycarbonylethoxymethyl)-methane is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PROTAC linker
  • Click chemistry reagent
  • Contains an azide group for CuAAc reaction
  • Can undergo SPAAC reactions with DBCO or BCN groups
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins