Learn More
Medchemexpress LLC Azido-PEG4-Amido-Tris | 1398044-55-7 | 96.4% | C15H30N4O8 | 100 MG
SDP
Catalog No. 5000355685
Encompass_Preferred
Azido-PEG4-Amido-Tris is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules possessing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs involve two distinct ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group for CuAAc reaction
  • Can undergo SPAAC reactions
  • Used in the synthesis of PROTACs
Catalog No. 50-003-55685 Supplier Medchemexpress LLC Supplier No. HY141249100MG
Azido-PEG4-Amido-Tris is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules possessing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs involve two distinct ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group for CuAAc reaction
  • Can undergo SPAAC reactions
  • Used in the synthesis of PROTACs