Learn More
Medchemexpress LLC Azido-PEG4-azide | 182760-73-2 | ≥97.0% | 288.30 | 250 MG
SDP
Catalog No. 5000348255
Encompass_Preferred
Azido-PEG4-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent features an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Click chemistry reagent
  • Contains an Azide group
  • Participates in CuAAc reactions with alkyne groups
  • Undergoes SPAAC reactions with DBCO or BCN groups
Catalog No. 50-003-48255 Supplier Medchemexpress LLC Supplier No. 250MGHY138692
Azido-PEG4-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent features an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Click chemistry reagent
  • Contains an Azide group
  • Participates in CuAAc reactions with alkyne groups
  • Undergoes SPAAC reactions with DBCO or BCN groups