Learn More
Medchemexpress LLC Azido-PEG4-beta-D-glucose | 1609083-15-9 | 99.9% | 381.38 | C14H27N3O9 | 5 MG
SDP
Catalog No. 5000223233
Encompass_Preferred
Azido-PEG4-beta-D-glucose is an azide-terminated PEG4 linker bearing a beta-D-glucose moiety, designed for use in click chemistry and PROTAC linker synthesis. It facilitates bioconjugation via copper-catalyzed and strain-promoted azide-alkyne cycloaddition and is supplied as a high-purity, well-characterized small-molecule linker for research applications.
  • Contains an azide functional group compatible with CuAAC and SPAAC reactions.
  • Incorporates a beta-D-glucose unit to increase hydrophilicity and potential targeting.
  • PEG4 spacer provides flexible linker length for conjugation strategies.
  • High purity suitable for research and bioconjugation workflows.
  • Supplied in small quantities for synthetic and analytical use.
Catalog No. 50-002-23233 Supplier Medchemexpress LLC Supplier No. HY1400125MG
Azido-PEG4-beta-D-glucose is an azide-terminated PEG4 linker bearing a beta-D-glucose moiety, designed for use in click chemistry and PROTAC linker synthesis. It facilitates bioconjugation via copper-catalyzed and strain-promoted azide-alkyne cycloaddition and is supplied as a high-purity, well-characterized small-molecule linker for research applications.
  • Contains an azide functional group compatible with CuAAC and SPAAC reactions.
  • Incorporates a beta-D-glucose unit to increase hydrophilicity and potential targeting.
  • PEG4 spacer provides flexible linker length for conjugation strategies.
  • High purity suitable for research and bioconjugation workflows.
  • Supplied in small quantities for synthetic and analytical use.