Learn More
Medchemexpress LLC Azido-PEG4-beta-D-glucose | 1609083-15-9 | 99.9% | C14H27N3O9 | 10MG
SDP
Catalog No. 5000260360
Encompass_Preferred
Azido-PEG4-beta-D-glucose is an azide-terminated PEG4 linker bearing a beta-D-glucose moiety, used as a click chemistry reagent and PROTAC linker. It enables efficient conjugation to alkyne-bearing molecules via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and to strained alkynes via strain-promoted alkyne-azide cycloaddition (SPAAC). The material is supplied at high purity for chemical biology and synthesis applications.
  • Azide-terminated PEG4 linker for efficient bioconjugation.
  • Beta-D-glucose unit enhances solubility and potential targeting.
  • Compatible with CuAAC and SPAAC click reactions.
  • High purity suitable for synthesis and analytical workflows.
  • Stable under sealed storage at -20°C; follow solvent storage guidelines.
Catalog No. 50-002-60360 Supplier Medchemexpress LLC Supplier No. HY14001210MG
Azido-PEG4-beta-D-glucose is an azide-terminated PEG4 linker bearing a beta-D-glucose moiety, used as a click chemistry reagent and PROTAC linker. It enables efficient conjugation to alkyne-bearing molecules via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and to strained alkynes via strain-promoted alkyne-azide cycloaddition (SPAAC). The material is supplied at high purity for chemical biology and synthesis applications.
  • Azide-terminated PEG4 linker for efficient bioconjugation.
  • Beta-D-glucose unit enhances solubility and potential targeting.
  • Compatible with CuAAC and SPAAC click reactions.
  • High purity suitable for synthesis and analytical workflows.
  • Stable under sealed storage at -20°C; follow solvent storage guidelines.