Learn More
Medchemexpress LLC Azido-PEG4-Boc | 581066-04-8 | 97% | C15H29N3O6 | 1 G
SDP
Catalog No. 5000353545
Encompass_Preferred
Azido-PEG4-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent and can exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-53545 Supplier Medchemexpress LLC Supplier No. HY1407771G
Azido-PEG4-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent and can exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups