Learn More
Medchemexpress LLC Azido-PEG4-(CH2)3OH | 2028281-87-8 | ≥97.0% | C11H23N3O5 | 250 MG
SDP
Catalog No. 5000352568
Encompass_Preferred
Azido-PEG4-(CH2)3OH is a PEG-based PROTAC linker designed for synthesizing PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling participation in various cycloaddition reactions. This product is suitable for laboratory research applications.
  • PEG-based PROTAC linker
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • Suitable for PROTAC synthesis
Catalog No. 50-003-52568 Supplier Medchemexpress LLC Supplier No. 250MGHY140809
Azido-PEG4-(CH2)3OH is a PEG-based PROTAC linker designed for synthesizing PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling participation in various cycloaddition reactions. This product is suitable for laboratory research applications.
  • PEG-based PROTAC linker
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • Suitable for PROTAC synthesis