Learn More
Medchemexpress LLC Azido-PEG4-CH2-Boc | 864681-04-9 | 97.0% | 333.38 g/mol | C14H27N3O6 | 5 G
SDP
Catalog No. 5000202331
Encompass_Preferred
Azido-PEG4-CH2-Boc is an azide-functionalized, Boc-protected polyethylene glycol (PEG4) linker used for bioconjugation and linker synthesis. It provides an azide handle for click chemistry and a protected amine for orthogonal functionalization, making it suitable for linker installation in medicinal chemistry and chemical biology workflows.
  • Provides an azide group for copper-catalyzed azide-alkyne cycloaddition
  • Contains a four-unit polyethylene glycol spacer to increase solubility
  • Features a boc-protected amine for orthogonal deprotection and coupling
  • Suitable for antibody-drug conjugate and PROTAC linker assembly
  • Available in multiple package sizes, including 5 g
Catalog No. 50-002-02331 Supplier Medchemexpress LLC Supplier No. HY426185G
Azido-PEG4-CH2-Boc is an azide-functionalized, Boc-protected polyethylene glycol (PEG4) linker used for bioconjugation and linker synthesis. It provides an azide handle for click chemistry and a protected amine for orthogonal functionalization, making it suitable for linker installation in medicinal chemistry and chemical biology workflows.
  • Provides an azide group for copper-catalyzed azide-alkyne cycloaddition
  • Contains a four-unit polyethylene glycol spacer to increase solubility
  • Features a boc-protected amine for orthogonal deprotection and coupling
  • Suitable for antibody-drug conjugate and PROTAC linker assembly
  • Available in multiple package sizes, including 5 g