Learn More
Medchemexpress LLC Azido-PEG4-NHS-ester | 1807534-82-2 | 96.0% | C14H22N4O8 | 25 MG
SDP
Catalog No. 5000352855
Encompass_Preferred
Azido-PEG4-NHS-ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent.
  • Functions as a click chemistry reagent
  • Contains an azide group for chemical reactions
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Viscous liquid appearance
  • Colorless to light yellow color
Catalog No. 50-003-52855 Supplier Medchemexpress LLC Supplier No. 25MGHY140765
Azido-PEG4-NHS-ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent.
  • Functions as a click chemistry reagent
  • Contains an azide group for chemical reactions
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Viscous liquid appearance
  • Colorless to light yellow color