Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-peg4-val-cit-pab-oh | 2055024-64-9 | MFCD29918229 | 98.0% | 652.74 g/mol | C29H48N8O9 | 100 MG
SDP

Catalog No. 5000202013
Encompass_Preferred

Azido-PEG4-Val-Cit-PAB-OH is a cleavable PEG-based linker used for assembling antibody-drug conjugates and other conjugates. It features an azide functional group for click chemistry and a protease-cleavable Val-Cit-PAB spacer that enables enzymatic payload release.

  • cleavable PEG-based linker for ADC and conjugate synthesis.
  • contains an azide group for CuAAC and SPAAC click reactions.
  • protease-sensitive Val-Cit-PAB spacer enabling payload release.
  • molecular weight 652.74 g/mol and formula C29H48N8O9.
  • high purity (98.0%) with recommended storage conditions for powder and solution.

Catalog No. 50-002-02013 Supplier Medchemexpress LLC Supplier No. HY140149100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG4-Val-Cit-PAB-OH is a cleavable PEG-based linker used for assembling antibody-drug conjugates and other conjugates. It features an azide functional group for click chemistry and a protease-cleavable Val-Cit-PAB spacer that enables enzymatic payload release.

  • cleavable PEG-based linker for ADC and conjugate synthesis.
  • contains an azide group for CuAAC and SPAAC click reactions.
  • protease-sensitive Val-Cit-PAB spacer enabling payload release.
  • molecular weight 652.74 g/mol and formula C29H48N8O9.
  • high purity (98.0%) with recommended storage conditions for powder and solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.