Learn More
Medchemexpress LLC Azido-PEG4-Val-Cit-PAB-OH | 2055024-64-9 | MFCD29918229 | 98.0% | 652.75 | C29H48N8O9 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14014950MG
Encompass_Preferred
Azido-PEG4-Val-Cit-PAB-OH is a cleavable four-unit PEG linker bearing an azide functional group and a protease-sensitive Val-Cit-PAB sequence, used for conjugation via click chemistry and for preparing antibody-drug conjugates or PROTACs.
  • cleavable PEG4 linker with azide functionality for click chemistry
  • protease-sensitive Val-Cit-PAB sequence for intracellular payload release
  • solid, white to off-white appearance suitable for storage as a powder
  • high purity suitable for research use
  • stable when stored at recommended low temperatures
Catalog No. 50-002-04145