Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG5-acid | 1425973-16-5 | >98.0% | 335.35 g/mol | C13H25N3O7 | 250 MG
SDP

Supplier:  Medchemexpress LLC HY130572250MG

Encompass_Preferred

Azido-PEG5-acid is an azide-functional, five-unit polyethylene glycol (PEG5) linker used for conjugation chemistry, including PROTAC and antibody-drug conjugate synthesis, and for click-chemistry applications. Supplied for research use only.

  • Peg5 linker with azide functionality for click chemistry.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted azide-alkyne cycloaddition (SPAAC) using DBCO or BCN reagents.
  • Purity ≥98.0%.
  • Appearance: colorless to light yellow liquid.
  • Storage: pure form stable at -20°C for 3 years and 4°C for 2 years.

Catalog No. 50-002-04029


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.