Learn More
Medchemexpress LLC Azido-PEG5-acid | 1425973-16-5 | MFCD23726612 | >98.0% | 335.35 g/mol | C13H25N3O7 | 100 MG
SDP
Catalog No. 5000202153
Encompass_Preferred
Azido-PEG5-acid is an azide-terminated polyethylene glycol (PEG5) linker designed for click chemistry and bioconjugation. It features a terminal carboxylic acid and an azide functional group that enable copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), making it useful for PROTAC construction, ADC linker assembly, and general spacer chemistry.
  • Azide-terminated PEG5 spacer for bioconjugation
  • Compatible with CuAAC and SPAAC click reactions
  • High purity for reliable coupling
  • Colorless to light yellow liquid, easy to handle
  • Stable under recommended storage conditions
Catalog No. 50-002-02153 Supplier Medchemexpress LLC Supplier No. HY130572100MG
Azido-PEG5-acid is an azide-terminated polyethylene glycol (PEG5) linker designed for click chemistry and bioconjugation. It features a terminal carboxylic acid and an azide functional group that enable copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), making it useful for PROTAC construction, ADC linker assembly, and general spacer chemistry.
  • Azide-terminated PEG5 spacer for bioconjugation
  • Compatible with CuAAC and SPAAC click reactions
  • High purity for reliable coupling
  • Colorless to light yellow liquid, easy to handle
  • Stable under recommended storage conditions