Learn More
Medchemexpress LLC Azido-PEG5-acid | 1425973-16-5 | MFCD23726612 | 98.0% | 335.35 g/mol | C13H25N3O7 | 500 MG
SDP
Catalog No. 5000204287
Encompass_Preferred
Azido-PEG5-acid is an azide-terminated, five-unit polyethylene glycol linker with a terminal carboxylic acid that enables click-chemistry bioconjugation reactions. It is used as a PROTAC and ADC linker and is compatible with copper-catalyzed azide-alkyne cycloaddition and strain-promoted cycloaddition reactions.
  • Azide functional group for CuAAC and SPAAC reactions.
  • Five-unit PEG spacer provides water solubility and flexibility.
  • Terminal carboxylic acid enables amide or ester coupling.
  • Typical purity ≥98.0%.
  • Suitable for linker synthesis and bioconjugation applications.
Catalog No. 50-002-04287 Supplier Medchemexpress LLC Supplier No. HY130572500MG
Azido-PEG5-acid is an azide-terminated, five-unit polyethylene glycol linker with a terminal carboxylic acid that enables click-chemistry bioconjugation reactions. It is used as a PROTAC and ADC linker and is compatible with copper-catalyzed azide-alkyne cycloaddition and strain-promoted cycloaddition reactions.
  • Azide functional group for CuAAC and SPAAC reactions.
  • Five-unit PEG spacer provides water solubility and flexibility.
  • Terminal carboxylic acid enables amide or ester coupling.
  • Typical purity ≥98.0%.
  • Suitable for linker synthesis and bioconjugation applications.