Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG5-alcohol | 86770-68-5 | ≥97.0% | 263.29 | C10H21N3O5 | 10 G
SDP

Catalog No. 5000202257
Encompass_Preferred

Azido-PEG5-alcohol is a polyethylene glycol (PEG) linker bearing a terminal azide and a terminal hydroxyl group. It functions as a non-cleavable, five-unit PEG spacer for bioconjugation and click chemistry applications.

  • Contains azide (-N3) and terminal alcohol (-OH) functional groups.
  • Water-soluble PEG linker suitable for click chemistry and PROTAC/ADC assembly.
  • Molecular weight ~263.29 g/mol; formula C10H21N3O5.
  • High purity, typically ≥97.0% by manufacturer specification.
  • Available in multiple pack sizes, including 10 g.

Catalog No. 50-002-02257 Supplier Medchemexpress LLC Supplier No. HY13021110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG5-alcohol is a polyethylene glycol (PEG) linker bearing a terminal azide and a terminal hydroxyl group. It functions as a non-cleavable, five-unit PEG spacer for bioconjugation and click chemistry applications.

  • Contains azide (-N3) and terminal alcohol (-OH) functional groups.
  • Water-soluble PEG linker suitable for click chemistry and PROTAC/ADC assembly.
  • Molecular weight ~263.29 g/mol; formula C10H21N3O5.
  • High purity, typically ≥97.0% by manufacturer specification.
  • Available in multiple pack sizes, including 10 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.