Learn More
Medchemexpress LLC Azido-PEG5-alcohol | 86770-68-5 | >97.0% | 263.29 g/mol | C10H21N3O5 | 25 G
SDP
Catalog No. 5000202138
Encompass_Preferred
Azido-PEG5-alcohol is an azide-terminated polyethylene glycol (PEG) linker containing five ethylene glycol units that is used as a click-chemistry reagent and as a linker in bioconjugation workflows. It participates in copper-catalyzed and strain-promoted azide-alkyne cycloaddition reactions and is supplied for research use only.
  • Azide functional group for click chemistry
  • Five-unit PEG spacer adds hydrophilicity and flexibility
  • Suitable for antibody-drug conjugate and PROTAC linker synthesis
  • Characterized by NMR and LC-MS, purity >97.0%
  • Available in multiple package sizes, including 25 G
Catalog No. 50-002-02138 Supplier Medchemexpress LLC Supplier No. HY13021125G
Azido-PEG5-alcohol is an azide-terminated polyethylene glycol (PEG) linker containing five ethylene glycol units that is used as a click-chemistry reagent and as a linker in bioconjugation workflows. It participates in copper-catalyzed and strain-promoted azide-alkyne cycloaddition reactions and is supplied for research use only.
  • Azide functional group for click chemistry
  • Five-unit PEG spacer adds hydrophilicity and flexibility
  • Suitable for antibody-drug conjugate and PROTAC linker synthesis
  • Characterized by NMR and LC-MS, purity >97.0%
  • Available in multiple package sizes, including 25 G