Learn More
Medchemexpress LLC Azido-PEG5-alcohol | 86770-68-5 | >97.0% | 263.29 g·mol⁻¹ | C10H21N3O5 | 5 G
SDP
Catalog No. 5000203146
Encompass_Preferred
Azido-PEG5-alcohol is an azide-terminated polyethylene glycol linker containing five ethylene glycol units and a terminal primary alcohol, used as a water-soluble reagent for bioconjugation and linker synthesis. It is commonly applied in click chemistry and in the construction of antibody-drug conjugates and PROTACs.
  • Azide functionality enables CuAAC and SPAAC click reactions.
  • Water-soluble PEG5 spacer improves solubility and flexibility.
  • Terminal primary alcohol provides a synthetic handle for derivatization.
  • Suitable for ADC linker synthesis and PROTAC construction.
  • Supplied as a solid with reported purity >97.0%.
Catalog No. 50-002-03146 Supplier Medchemexpress LLC Supplier No. HY1302115G
Azido-PEG5-alcohol is an azide-terminated polyethylene glycol linker containing five ethylene glycol units and a terminal primary alcohol, used as a water-soluble reagent for bioconjugation and linker synthesis. It is commonly applied in click chemistry and in the construction of antibody-drug conjugates and PROTACs.
  • Azide functionality enables CuAAC and SPAAC click reactions.
  • Water-soluble PEG5 spacer improves solubility and flexibility.
  • Terminal primary alcohol provides a synthetic handle for derivatization.
  • Suitable for ADC linker synthesis and PROTAC construction.
  • Supplied as a solid with reported purity >97.0%.