Learn More
Medchemexpress LLC Azido-PEG5-amine | 516493-93-9 | 98.0% | C12H26N4O5 | 250 MG
SDP
Catalog No. 5000352583
Encompass_Preferred
Azido-PEG5-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Peg-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-52583 Supplier Medchemexpress LLC Supplier No. 250MGHY140214
Azido-PEG5-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Peg-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups