Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG5-CH2CO2H | 217180-81-9 | 98.9% | 321.33 g/mol | C12H23N3O7 | 100 MG
SDP

Catalog No. 5000202970
Encompass_Preferred

Azido-PEG5-CH2CO2H is a cleavable, azide-functional polyethylene glycol (PEG) linker containing five ethylene glycol units. It is intended for use in antibody-drug conjugate (ADC) and PROTAC synthesis, and it functions as a click-chemistry reagent for copper-catalyzed and strain-promoted azide-alkyne cycloadditions.

  • Cleavable PEG5 linker suitable for ADC and PROTAC synthesis.
  • Azide functional group for copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • Molecular weight 321.33 g/mol and formula C12H23N3O7.
  • Purity 98.9%.
  • Appearance: liquid; store protected from light at -20°C; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Common packaging: 25 mg, 50 mg, 100 mg, 250 mg, 500 mg, 1 g.

Catalog No. 50-002-02970 Supplier Medchemexpress LLC Supplier No. HY130194100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG5-CH2CO2H is a cleavable, azide-functional polyethylene glycol (PEG) linker containing five ethylene glycol units. It is intended for use in antibody-drug conjugate (ADC) and PROTAC synthesis, and it functions as a click-chemistry reagent for copper-catalyzed and strain-promoted azide-alkyne cycloadditions.

  • Cleavable PEG5 linker suitable for ADC and PROTAC synthesis.
  • Azide functional group for copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • Molecular weight 321.33 g/mol and formula C12H23N3O7.
  • Purity 98.9%.
  • Appearance: liquid; store protected from light at -20°C; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Common packaging: 25 mg, 50 mg, 100 mg, 250 mg, 500 mg, 1 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.