Learn More
Medchemexpress LLC Azido-PEG5-CH2CO2H | 217180-81-9 | 98.9% | 321.33 g/mol | C12H23N3O7 | 250 MG
SDP
Catalog No. 5000202259
Encompass_Preferred
Azido-PEG5-CH2CO2H is a cleavable, azide-functionalized polyethylene glycol (PEG) linker with a terminal carboxylic acid that provides a hydrophilic spacer and a reactive azide handle for bioconjugation and linker synthesis.
  • Cleavable 5-unit PEG linker suitable for linker-based conjugation.
  • Azide functional group for copper-catalyzed or strain-promoted click reactions.
  • Terminal carboxylic acid enables further derivatization or coupling.
  • Hydrophilic spacer increases solubility in aqueous media.
  • High purity suitable for chemical synthesis and bioconjugation.
  • Available in 250 MG pack size.
Catalog No. 50-002-02259 Supplier Medchemexpress LLC Supplier No. HY130194250MG
Azido-PEG5-CH2CO2H is a cleavable, azide-functionalized polyethylene glycol (PEG) linker with a terminal carboxylic acid that provides a hydrophilic spacer and a reactive azide handle for bioconjugation and linker synthesis.
  • Cleavable 5-unit PEG linker suitable for linker-based conjugation.
  • Azide functional group for copper-catalyzed or strain-promoted click reactions.
  • Terminal carboxylic acid enables further derivatization or coupling.
  • Hydrophilic spacer increases solubility in aqueous media.
  • High purity suitable for chemical synthesis and bioconjugation.
  • Available in 250 MG pack size.