Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG5-CH2CO2H | 217180-81-9 | 98.9% | 321.33 g/mol | C12H23N3O7 | 50 MG
SDP

Catalog No. 5000202381
Encompass_Preferred

Azido-PEG5-CH2CO2H is an azido-terminated PEG5 linker with a carboxylic acid terminus used as a cleavable linker in antibody-drug conjugate (ADC) and PROTAC synthesis. The compound is supplied as a liquid with high purity and specific formulation and storage recommendations.

  • Cleavable five-unit PEG linker.
  • Terminal azide and carboxylic acid functionality.
  • Purity: 98.9%.
  • Molecular weight: 321.33 g/mol.
  • Soluble in DMSO (100 mg/mL); may require sonication and fresh DMSO.
  • In vivo formulation protocols report ≥2.5 mg/mL solubility using recommended vehicles.
  • Storage: -20°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-02381 Supplier Medchemexpress LLC Supplier No. HY13019450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG5-CH2CO2H is an azido-terminated PEG5 linker with a carboxylic acid terminus used as a cleavable linker in antibody-drug conjugate (ADC) and PROTAC synthesis. The compound is supplied as a liquid with high purity and specific formulation and storage recommendations.

  • Cleavable five-unit PEG linker.
  • Terminal azide and carboxylic acid functionality.
  • Purity: 98.9%.
  • Molecular weight: 321.33 g/mol.
  • Soluble in DMSO (100 mg/mL); may require sonication and fresh DMSO.
  • In vivo formulation protocols report ≥2.5 mg/mL solubility using recommended vehicles.
  • Storage: -20°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.