Learn More
Medchemexpress LLC Azido-PEG5-NHS ester | 1433996-86-1 | ≥98.0% | C17H28N4O9 | 50 MG
SDP
Catalog No. 5000353158
Encompass_Preferred
Azido-PEG5-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound functions as a click chemistry reagent due to its azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It also facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups, leveraging the ubiquitin-proteasome system for selective protein degradation.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Enables targeted protein degradation
Catalog No. 50-003-53158 Supplier Medchemexpress LLC Supplier No. HY14075750MG
Azido-PEG5-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound functions as a click chemistry reagent due to its azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It also facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups, leveraging the ubiquitin-proteasome system for selective protein degradation.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Enables targeted protein degradation