Learn More
Medchemexpress LLC Azido-PEG5-triethoxysilane | 3025193-22-7 | 95.0% | C22H46N4O9Si | 50 MG
SDP
Catalog No. 5000354995
Encompass_Preferred
Azido-PEG5-triethoxysilane is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent containing an Azide group, enabling it to participate in various cycloaddition reactions. PROTACs are designed to degrade target proteins via the intracellular ubiquitin-proteasome system.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-54995 Supplier Medchemexpress LLC Supplier No. HY14085650MG
Azido-PEG5-triethoxysilane is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent containing an Azide group, enabling it to participate in various cycloaddition reactions. PROTACs are designed to degrade target proteins via the intracellular ubiquitin-proteasome system.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups