Learn More
Medchemexpress LLC Azido-PEG5-triethoxysilane | 3025193-22-7 | MFCD31693730 | 95.0% | C22H46N4O9Si | 25 MG
SDP
Catalog No. 5000357374
Encompass_Preferred
Azido-PEG5-triethoxysilane is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, which allows it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains an Azide group for copper-catalyzed azide-alkyne cycloaddition reactions.
  • Can undergo strain-promoted alkyne-azide cycloaddition reactions.
  • PROTACs enable selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
Catalog No. 50-003-57374 Supplier Medchemexpress LLC Supplier No. 25MGHY140856
Azido-PEG5-triethoxysilane is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, which allows it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Used in the synthesis of PROTACs.
  • Functions as a click chemistry reagent.
  • Contains an Azide group for copper-catalyzed azide-alkyne cycloaddition reactions.
  • Can undergo strain-promoted alkyne-azide cycloaddition reactions.
  • PROTACs enable selective degradation of target proteins via the intracellular ubiquitin-proteasome system.