Learn More
Medchemexpress LLC Azido-PEG6-acid | 361189-66-4 | 99.8% | C15H29N3O8 | 100 MG
SDP
Catalog No. 5000356641
Encompass_Preferred
Azido-PEG6-acid is a PEG-based PROTAC linker used in PROTAC synthesis. It is a click chemistry reagent containing an Azide group, which can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne groups. It also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group for CuAAc reactions with alkyne groups
  • Undergoes SPAAC reactions with DBCO or BCN groups
  • Leverages the intracellular ubiquitin-proteasome system for selective degradation of target proteins
Catalog No. 50-003-56641 Supplier Medchemexpress LLC Supplier No. HY140453100MG
Azido-PEG6-acid is a PEG-based PROTAC linker used in PROTAC synthesis. It is a click chemistry reagent containing an Azide group, which can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne groups. It also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group for CuAAc reactions with alkyne groups
  • Undergoes SPAAC reactions with DBCO or BCN groups
  • Leverages the intracellular ubiquitin-proteasome system for selective degradation of target proteins