Learn More
Medchemexpress LLC Azido-PEG6-alcohol | 86770-69-6 | 98.0% | 10 G
SDP
Catalog No. 5000340838
Encompass_Preferred
Azido-PEG6-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It also serves as a non-cleavable 6-unit PEG ADC linker for synthesizing antibody-drug conjugates (ADCs). This compound functions as a click chemistry reagent, containing an Azide group, and can undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Peg-based PROTAC linker.
  • Non-cleavable 6-unit PEG ADC linker.
  • Click chemistry reagent.
  • Contains an azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
Catalog No. 50-003-40838 Supplier Medchemexpress LLC Supplier No. HY13053710G
Azido-PEG6-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It also serves as a non-cleavable 6-unit PEG ADC linker for synthesizing antibody-drug conjugates (ADCs). This compound functions as a click chemistry reagent, containing an Azide group, and can undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Peg-based PROTAC linker.
  • Non-cleavable 6-unit PEG ADC linker.
  • Click chemistry reagent.
  • Contains an azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.