Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG6-alcohol | 86770-69-6 | MFCD22056309 | 98.0% | 307.34 g/mol | C12H25N3O6 | 5 G
SDP

Catalog No. 5000204265
Encompass_Preferred

Azido-PEG6-alcohol is an azide-terminated polyethylene glycol (PEG6) linker reagent supplied as a 5 g reagent for use in click chemistry and conjugation workflows. It contains a terminal azide and a terminal hydroxyl group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.

  • Azide-terminated PEG6 linker suitable for conjugation chemistry.
  • Terminal hydroxyl group enables further functionalization.
  • Compatible with CuAAC and SPAAC click reactions.
  • Purity 98.0%.
  • Colorless to light yellow liquid.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-04265 Supplier Medchemexpress LLC Supplier No. HY1305375G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG6-alcohol is an azide-terminated polyethylene glycol (PEG6) linker reagent supplied as a 5 g reagent for use in click chemistry and conjugation workflows. It contains a terminal azide and a terminal hydroxyl group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.

  • Azide-terminated PEG6 linker suitable for conjugation chemistry.
  • Terminal hydroxyl group enables further functionalization.
  • Compatible with CuAAC and SPAAC click reactions.
  • Purity 98.0%.
  • Colorless to light yellow liquid.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.