Learn More
Medchemexpress LLC Azido-PEG6-azide | 1243536-56-2 | 96.2% | 376.41 g/mol | C14H28N6O6 | 5 MG
SDP
Catalog No. 5000262407
Encompass_Preferred
Azido-PEG6-azide is a bifunctional PEG6 linker bearing azide groups at both termini, designed for click-chemistry conjugation and linker design in small-molecule and biomolecule synthesis. It is supplied for research use and provided in small, precisely measured packs for laboratory applications.
  • Bifunctional PEG6 linker with azide groups at both ends.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioconjugation.
  • Suitable for PROTAC linker synthesis and small-molecule conjugation.
  • High purity (~96.2%) for reproducible synthetic results.
  • Molecular weight 376.41 g/mol; formula C14H28N6O6.
  • Available in small research packs, including a 5 mg option.
  • Colorless to light yellow liquid; store sealed, away from moisture.
Catalog No. 50-002-62407 Supplier Medchemexpress LLC Supplier No. HY1333935MG
Azido-PEG6-azide is a bifunctional PEG6 linker bearing azide groups at both termini, designed for click-chemistry conjugation and linker design in small-molecule and biomolecule synthesis. It is supplied for research use and provided in small, precisely measured packs for laboratory applications.
  • Bifunctional PEG6 linker with azide groups at both ends.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioconjugation.
  • Suitable for PROTAC linker synthesis and small-molecule conjugation.
  • High purity (~96.2%) for reproducible synthetic results.
  • Molecular weight 376.41 g/mol; formula C14H28N6O6.
  • Available in small research packs, including a 5 mg option.
  • Colorless to light yellow liquid; store sealed, away from moisture.