Learn More
Medchemexpress LLC Azido-PEG6-C2-Boc | 406213-76-1 | >98% | C19H37N3O8 | 50 MG
SDP
Catalog No. 5000355697
Encompass_Preferred
Azido-PEG6-C2-Boc is a PEG-based PROTAC linker, primarily used in the synthesis of PROTACs. It also functions as a versatile click chemistry reagent, enabling various bioconjugation reactions.
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne-containing molecules
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • Functions as a reference compound
Catalog No. 50-003-55697 Supplier Medchemexpress LLC Supplier No. HY14077950MG
Azido-PEG6-C2-Boc is a PEG-based PROTAC linker, primarily used in the synthesis of PROTACs. It also functions as a versatile click chemistry reagent, enabling various bioconjugation reactions.
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne-containing molecules
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • Functions as a reference compound