Learn More
Medchemexpress LLC Azido-PEG6-CH2COOH | 880129-82-8 | ≥98.0% | 365.38 | 5 G
SDP
Catalog No. 5000364467
Encompass_Preferred
Azido-PEG6-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Suitable for synthesizing PROTACs
  • Click chemistry reagent due to azide group
  • Enables CuAAc reactions
  • Participates in SPAAC reactions with DBCO or BCN groups
Catalog No. 50-003-64467 Supplier Medchemexpress LLC Supplier No. 5GHY134696
Azido-PEG6-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Suitable for synthesizing PROTACs
  • Click chemistry reagent due to azide group
  • Enables CuAAc reactions
  • Participates in SPAAC reactions with DBCO or BCN groups