Learn More
Medchemexpress LLC Azido-PEG6-MS | 352439-38-4 | MFCD34471264 | >95.0% | 385.43 | C13H27N3O8S | 5 MG
SDP
Catalog No. 5000223388
Encompass_Preferred
Azido-PEG6-MS is an azide-terminated PEG6 linker used as a click-chemistry reagent and in PROTAC linker synthesis. It provides a short, flexible, hydrophilic spacer with a terminal azide that enables copper-catalyzed or strain-promoted azide-alkyne cycloaddition for attaching small molecules or biomolecules in medicinal chemistry and bioconjugation applications.
  • Contains an azide functional group for click chemistry reactions.
  • Provides a PEG6 spacer for hydrophilicity and conformational flexibility.
  • Suitable for PROTAC linker synthesis and bioconjugation workflows.
  • Supplied in small-scale quantities appropriate for research use.
  • Compatible with common conjugation conditions, including CuAAC.
Catalog No. 50-002-23388 Supplier Medchemexpress LLC Supplier No. HY1383635MG
Azido-PEG6-MS is an azide-terminated PEG6 linker used as a click-chemistry reagent and in PROTAC linker synthesis. It provides a short, flexible, hydrophilic spacer with a terminal azide that enables copper-catalyzed or strain-promoted azide-alkyne cycloaddition for attaching small molecules or biomolecules in medicinal chemistry and bioconjugation applications.
  • Contains an azide functional group for click chemistry reactions.
  • Provides a PEG6 spacer for hydrophilicity and conformational flexibility.
  • Suitable for PROTAC linker synthesis and bioconjugation workflows.
  • Supplied in small-scale quantities appropriate for research use.
  • Compatible with common conjugation conditions, including CuAAC.