Learn More
Medchemexpress LLC Azido-PEG6-NHS ester | 2055014-64-5 | 98.0% | 1 G
SDP
Catalog No. 5000338303
Encompass_Preferred
Azido-PEG6-NHS ester is a cleavable 6 unit PEG ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs). It also functions as a PEG- and Alkyl/ether based PROTAC linker for synthesizing PROTACs. This compound is a click chemistry reagent, featuring an Azide group, which allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Cleavable linker
  • PEG linker type
  • Alkyl/ether linker type
  • Click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition reactions
  • Used in ADC synthesis
  • Used in PROTAC synthesis
Catalog No. 50-003-38303 Supplier Medchemexpress LLC Supplier No. HY1304741G
Azido-PEG6-NHS ester is a cleavable 6 unit PEG ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs). It also functions as a PEG- and Alkyl/ether based PROTAC linker for synthesizing PROTACs. This compound is a click chemistry reagent, featuring an Azide group, which allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Cleavable linker
  • PEG linker type
  • Alkyl/ether linker type
  • Click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition reactions
  • Used in ADC synthesis
  • Used in PROTAC synthesis