Learn More
Medchemexpress LLC Azido-PEG6-NHS ester | 2055014-64-5 | 98.0% | 250 MG
SDP
Catalog No. 5000341967
Encompass_Preferred
Azido-PEG6-NHS ester is a cleavable PEG ADC linker. It is also a PEG- and alkyl/ether-based PROTAC linker. This compound is a click chemistry reagent, featuring an azide group for copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Cleavable PEG ADC linker
  • Used in antibody-drug conjugate (ADC) synthesis
  • PEG- and alkyl/ether-based PROTAC linker
  • Suitable for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-41967 Supplier Medchemexpress LLC Supplier No. 250MGHY130474
Azido-PEG6-NHS ester is a cleavable PEG ADC linker. It is also a PEG- and alkyl/ether-based PROTAC linker. This compound is a click chemistry reagent, featuring an azide group for copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Cleavable PEG ADC linker
  • Used in antibody-drug conjugate (ADC) synthesis
  • PEG- and alkyl/ether-based PROTAC linker
  • Suitable for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups