Learn More
Medchemexpress LLC Azido-PEG6-NHS ester | 2055014-64-5 | 98.0% | 476.48 g/mol | C19H32N4O10 | 100 MG
SDP
Catalog No. 5000201900
Encompass_Preferred
Azido-PEG6-NHS ester is a bifunctional PEG-based linker containing an azide group and an N-hydroxysuccinimide (NHS) ester; it is used as a cleavable six-unit PEG linker for antibody-drug conjugate and PROTAC synthesis, enabling combined click chemistry and amine-reactive coupling.
  • Bifunctional linker with azide and NHS ester.
  • Six-unit PEG spacer provides flexibility and solubility.
  • Enables click chemistry (azide-alkyne) and NHS amine coupling.
  • Suitable for ADC and PROTAC linker synthesis.
  • Available in multiple package sizes, including 100 MG.
Catalog No. 50-002-01900 Supplier Medchemexpress LLC Supplier No. HY130474100MG
Azido-PEG6-NHS ester is a bifunctional PEG-based linker containing an azide group and an N-hydroxysuccinimide (NHS) ester; it is used as a cleavable six-unit PEG linker for antibody-drug conjugate and PROTAC synthesis, enabling combined click chemistry and amine-reactive coupling.
  • Bifunctional linker with azide and NHS ester.
  • Six-unit PEG spacer provides flexibility and solubility.
  • Enables click chemistry (azide-alkyne) and NHS amine coupling.
  • Suitable for ADC and PROTAC linker synthesis.
  • Available in multiple package sizes, including 100 MG.