Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG6-NHS ester | 2055014-64-5 | 98.0% | 476.48 g/mol | C19H32N4O10 | 50 MG
SDP

Catalog No. 5000203381
Encompass_Preferred

Azido-PEG6-NHS ester is a cleavable six-unit polyethylene glycol (PEG) linker containing an azide and an N-hydroxysuccinimide (NHS) ester. It is used as a click-chemistry reagent for conjugation reactions and as a linker in antibody-drug conjugate and PROTAC synthesis.

  • Cleavable six-unit PEG linker for bioconjugation.
  • Azide group enables CuAAC and SPAAC click reactions.
  • NHS ester reacts with primary amines for efficient coupling.
  • Molecular weight 476.48 g/mol, formula C19H32N4O10.
  • Purity 98.0% and liquid form, colorless to light yellow.
  • Store pure material at -20°C; in solvent store at -80°C or -20°C as specified.
  • Available in small quantities such as 50 mg for lab-scale use.

Catalog No. 50-002-03381 Supplier Medchemexpress LLC Supplier No. HY13047450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG6-NHS ester is a cleavable six-unit polyethylene glycol (PEG) linker containing an azide and an N-hydroxysuccinimide (NHS) ester. It is used as a click-chemistry reagent for conjugation reactions and as a linker in antibody-drug conjugate and PROTAC synthesis.

  • Cleavable six-unit PEG linker for bioconjugation.
  • Azide group enables CuAAC and SPAAC click reactions.
  • NHS ester reacts with primary amines for efficient coupling.
  • Molecular weight 476.48 g/mol, formula C19H32N4O10.
  • Purity 98.0% and liquid form, colorless to light yellow.
  • Store pure material at -20°C; in solvent store at -80°C or -20°C as specified.
  • Available in small quantities such as 50 mg for lab-scale use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.